Assay Details
Gene Symbol: Nsun2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145354.5 | NP_663329.3 | 9 - 10 | 1102 | 67 | |
GenBank mRNA | AK030124.1 | - | 8 - 9 | 987 | 67 | |
AK150631.1 | - | 9 - 10 | 1056 | 67 | ||
AK151917.1 | - | 9 - 10 | 1056 | 67 | ||
AK154789.1 | - | 9 - 10 | 1094 | 67 | ||
AK171399.1 | - | 9 - 10 | 1059 | 67 | ||
BC013625.1 | - | 5 - 6 | 574 | 4011674 | 67 | |
BC023735.1 | - | 3 - 4 | 371 | 67 | ||
BC025549.1 | - | 9 - 10 | 1066 | 67 | ||
DQ490066.1 | - | 9 - 10 | 1065 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
