Assay Details
Gene Symbol: Slc22a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144785.2 | NP_659034.1 | 8 - 9 | 1512 | 103 | |
GenBank mRNA | AF529221.1 | - | 8 - 9 | 1441 | 103 | |
BC021384.1 | - | 8 - 9 | 1478 | 5064542 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057370 | BC021384.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057370 | NM_144785.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057370 | AF529221.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334966 | AK086468.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334966 | AY313449.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334966 | BC082538.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334966 | NM_177161.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334966 | XM_001472921.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260181 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185399 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31859 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 464166 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821244 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876327 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 273705 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15947 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644977 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724652 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141845 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831375 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496121 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514772 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359864 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506972 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135790 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89927 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750627 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90446 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 66381 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 158103 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156696 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121292 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172020 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057370 | BC021384.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057370 | NM_144785.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057370 | AF529221.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941047 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941040 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941042 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941033 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941044 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941035 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941051 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941034 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941046 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941038 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941037 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941049 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941036 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941048 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31941043 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09099337 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09099324 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121292 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172020 | - | - |
Whole Mouse Genome G4122A | A_52_P94633 | BC021384.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P94633 | AK052752.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P94633 | NM_144785.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P94633 | AF529221.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104967 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172020 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121292 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14171607 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14301682 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14171604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31941018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31940958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14462062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14163511 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14163518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941025 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940976 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14277632 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941029 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941018 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941043 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14462063 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941044 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941024 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14167038 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14301682 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941026 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940875 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941012 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941049 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940978 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14277638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31941034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14277640 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14171609 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31940963 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10465718 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465723 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465719 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465722 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465720 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465724 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465717 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465716 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465725 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10465721 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425038_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10465715 | - | - |
Mouse Genome 430 2.0 Array | 1425038_at | - | - |
Mouse Genome 430 2.0 Array | 1425038_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 207151_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 207151_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1900009838.mm.1 | TC1900001053.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900009838.mm.1 | NM_144785.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900009838.mm.1 | NM_144785.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900009837.mm.1 | TC1900001053.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900009837.mm.1 | NM_144785.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900009837.mm.1 | NM_144785.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900009839.mm.1 | TC1900001053.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1900009839.mm.1 | NM_144785.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1900009839.mm.1 | NM_144785.2 | Refseq |
Mouse Expression Set 430 | 1425038_at | - | - |
Mouse Expression Set 430 | 1425038_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425038_at | - | - |
Mouse Genome 430A 2.0 Array | 1425038_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425038_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425038_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 629982 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2810546 | - | - |
MouseWG-6 v2 Expression BeadChip | 4280670 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5670541 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2810546 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106980619 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4280670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002810546 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004280670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005670541 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2652817 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2652112 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2591076 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2572143 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2591076 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2652112 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2652817 | - | - |
mouse-6 v1.1 (Illumina) | scl52883.10.1_185-S | - | - |
mouse-6 v1.1 (Illumina) | scl000427.1_81-S | - | - |
mouse-6 v1.1 (Illumina) | scl000504.1_48-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106980619 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2810546 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4280670 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5670541 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl000427.1_81-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl52883.10.1_185-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl000504.1_48-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5670541 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280670 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106980619 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2810546 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2800740 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2652112 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2591076 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2800732 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2572143 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2591076 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2800732 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2652112 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2800740 | - | - |
