Assay Details
Gene Symbol: Slc24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144813.1 | NP_659062.1 | 7 - 8 | 3145 | 70 | |
GenBank mRNA | BC016094.1 | - | 7 - 8 | 3145 | 4504496 | 70 |
BC044859.1 | - | 6 - 7 | 2581 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
