Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100019888.mm.1 |
TC0100002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019888.mm.1 |
NM_001160038.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019888.mm.1 |
NM_001160038.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019888.mm.1 |
NM_001160039.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019888.mm.1 |
NM_001160039.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019888.mm.1 |
NM_001160040.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019888.mm.1 |
NM_001160040.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019888.mm.1 |
NM_145518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019888.mm.1 |
NM_145518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019889.mm.1 |
TC0100002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019889.mm.1 |
NM_001160038.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019889.mm.1 |
NM_001160038.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019889.mm.1 |
NM_001160039.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019889.mm.1 |
NM_001160039.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019889.mm.1 |
NM_001160040.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019889.mm.1 |
NM_001160040.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019889.mm.1 |
NM_145518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019889.mm.1 |
NM_145518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019890.mm.1 |
TC0100002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019890.mm.1 |
NM_001160038.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019890.mm.1 |
NM_001160038.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019890.mm.1 |
NM_001160039.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019890.mm.1 |
NM_001160039.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019890.mm.1 |
NM_001160040.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019890.mm.1 |
NM_001160040.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019890.mm.1 |
NM_145518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019890.mm.1 |
NM_145518.2 |
Refseq |