Assay Details
Gene Symbol: Cwh43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181323.2 | NP_851840.1 | 12 - 13 | 1938 | 74 | |
GenBank mRNA | AK035550.1 | - | 11 - 12 | 1531 | 74 | |
AK081976.1 | - | 12 - 13 | 1938 | 74 | ||
BC016523.1 | - | 12 - 13 | 1766 | 3584006 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
