Assay Details
Gene Symbol: Cwh43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181323.2 | NP_851840.1 | 14 - 15 | 2144 | 114 | |
GenBank mRNA | AK035550.1 | - | 13 - 14 | 1737 | 114 | |
AK081976.1 | - | 14 - 15 | 2144 | 114 | ||
BC016523.1 | - | 14 - 15 | 1972 | 3584006 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
