Assay Details
Gene Symbol: Acbd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102436.1 | NP_001095906.1 | 7 - 8 | 891 | 65 | |
NM_001102437.1 | NP_001095907.1 | 7 - 8 | 1018 | 65 | ||
NM_001102438.1 | NP_001095908.1 | 7 - 8 | 1018 | 65 | ||
NM_028793.3 | NP_083069.1 | 7 - 8 | 891 | 65 | ||
GenBank mRNA | AK005001.2 | - | 6 - 7 | 807 | 65 | |
AK050450.1 | - | 7 - 8 | 857 | 65 | ||
AK147839.1 | - | 7 - 8 | 1018 | 65 | ||
BC035202.1 | - | 7 - 8 | 947 | 65 | ||
BC053518.1 | - | 7 - 8 | 985 | 65 | ||
BC061484.1 | - | 6 - 7 | 789 | 5355466 | 65 | |
BC084584.1 | - | 7 - 8 | 823 | 5257773 | 65 | |
BC090641.1 | - | 7 - 8 | 842 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
