Assay Details
Gene Symbol: Dpp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028906.2 | NP_083182.2 | 14 - 15 | 2006 | 82 | |
GenBank mRNA | AK016546.1 | - | 14 - 15 | 2045 | 82 | |
AK029788.1 | - | 12 - 13 | 3877 | 82 | ||
AK155605.1 | - | 14 - 15 | 2051 | 82 | ||
AK171895.1 | - | 1 - 2 | 76 | 82 | ||
BC043124.1 | - | 14 - 15 | 2006 | 6410075 | 82 | |
BC059222.1 | - | 14 - 15 | 2006 | 6410075 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
