Assay Details
Gene Symbol: Aldh16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145954.1 | NP_666066.1 | 9 - 10 | 1253 | 63 | |
GenBank mRNA | AK030673.1 | - | 9 - 10 | 1246 | 63 | |
AK144967.1 | - | 1252 | 63 | |||
AK144995.1 | - | 9 - 10 | 1260 | 63 | ||
AK151691.1 | - | 9 - 10 | 1253 | 63 | ||
AK161198.1 | - | 9 - 10 | 1291 | 63 | ||
AK164374.1 | - | 9 - 10 | 1299 | 63 | ||
AK169346.1 | - | 1604 | 63 | |||
AK170276.1 | - | 9 - 10 | 1292 | 63 | ||
AK220200.1 | - | 9 - 10 | 1229 | 63 | ||
BC013548.1 | - | 9 - 10 | 1253 | 4222058 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
