Assay Details
Gene Symbol: Thoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172438.3 | NP_766026.1 | 4 - 5 | 406 | 82 | |
GenBank mRNA | AK050734.1 | - | 4 - 5 | 406 | 82 | |
AK173078.1 | - | 2 - 3 | 244 | 82 | ||
AK207100.1 | - | 1 - 2 | 72 | 82 | ||
BC021779.1 | - | 3 - 4 | 380 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
