Assay Details
Gene Symbol: Rrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163734.1 | NP_001157206.1 | 11 - 12 | 1914 | 59 | |
NM_028244.2 | NP_082520.2 | 14 - 15 | 2230 | 59 | ||
GenBank mRNA | AK129078.1 | - | 11 - 12 | 1914 | 59 | |
BC016569.1 | - | 14 - 15 | 2230 | 3157173 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
