Assay Details
Gene Symbol: Tdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028354.4 | NP_082630.2 | 9 - 10 | 1181 | 119 | |
GenBank mRNA | AK013427.1 | - | 8 - 9 | 656 | 119 | |
AK014855.1 | - | 9 - 10 | 1161 | 119 | ||
AK033868.1 | - | 9 - 10 | 1179 | 119 | ||
AK189362.1 | - | 3 - 4 | 298 | 119 | ||
BC019804.1 | - | 6 - 7 | 505 | 4018534 | 119 | |
BC065162.1 | - | 8 - 9 | 1151 | 6402921 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
