Assay Details
Gene Symbol: Nln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029447.2 | NP_083723.1 | 6 - 7 | 977 | 72 | |
GenBank mRNA | AK079090.1 | - | 6 - 7 | 978 | 72 | |
AK138399.1 | - | 6 - 7 | 950 | 72 | ||
AK146364.1 | - | 6 - 7 | 926 | 72 | ||
AK170508.1 | - | 6 - 7 | 978 | 72 | ||
AK171432.1 | - | 6 - 7 | 978 | 72 | ||
BC016224.1 | - | 6 - 7 | 950 | 4486941 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
