Assay Details
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012518.3 | NP_001012536.2 | 12 - 13 | 2053 | 70 | |
NM_001109686.2 | NP_001103156.1 | 11 - 12 | 1894 | 70 | ||
NM_001109687.2 | NP_001103157.1 | 11 - 12 | 1909 | 70 | ||
NM_172545.4 | NP_766133.2 | 12 - 13 | 2032 | 70 | ||
GenBank mRNA | AB205007.1 | - | 12 - 13 | 2059 | 70 | |
AK049454.1 | - | 9 - 10 | 1303 | 70 | ||
AK052174.1 | - | 12 - 13 | 2016 | 70 | ||
AK054555.1 | - | 12 - 13 | 2001 | 70 | ||
AK082062.1 | - | 12 - 13 | 2043 | 70 | ||
BC056938.1 | - | 9 - 10 | 1610 | 70 | ||
BC089302.1 | - | 9 - 10 | 1762 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
