Assay Details
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172560.3 | NP_766148.2 | 9 - 10 | 2204 | 56 | |
GenBank mRNA | AK036710.1 | - | 8 - 9 | 2024 | 56 | |
AK147774.1 | - | 9 - 10 | 2090 | 56 | ||
BC058512.1 | - | 9 - 10 | 1037 | 6817014 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
