Assay Details
Gene Symbol: Tlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172664.3 | NP_766252.2 | 15 - 16 | 1740 | 88 | |
GenBank mRNA | AK029773.1 | - | 14 - 15 | 1466 | 88 | |
AK172897.1 | - | 9 - 10 | 1004 | 88 | ||
BC051641.1 | - | 15 - 16 | 1740 | 6507968 | 88 | |
BC057369.1 | - | 17 - 18 | 1979 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
