Assay Details
Gene Symbol: Plekha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040271.1 | - | 22 - 23 | 3338 | 111 | |
AK146866.1 | - | 3529 | 111 | |||
BC060678.1 | - | 21 - 22 | 3259 | 111 | ||
EU380771.1 | - | 3555 | 111 | |||
EU380772.1 | - | 3613 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
