Assay Details
Gene Symbol: Arhgap28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172964.4 | NP_766552.3 | 8 - 9 | 1232 | 62 | |
GenBank mRNA | AK087523.1 | - | 8 - 9 | 1245 | 62 | |
AK133720.1 | - | 8 - 9 | 1074 | 62 | ||
AK141901.1 | - | 8 - 9 | 1182 | 62 | ||
AK162051.1 | - | 2 - 3 | 272 | 62 | ||
AK173147.1 | - | 7 - 8 | 859 | 62 | ||
BC066788.1 | - | 8 - 9 | 1222 | 30049271 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
