Assay Details
Gene Symbol: Pmpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173180.3 | NP_775272.1 | 12 - 13 | 1425 | 69 | |
GenBank mRNA | AK004549.1 | - | 12 - 13 | 1425 | 69 | |
AK075965.1 | - | 12 - 13 | 1425 | 69 | ||
AK147110.1 | - | 12 - 13 | 1428 | 69 | ||
AK158990.1 | - | 12 - 13 | 1393 | 69 | ||
AK161291.1 | - | 12 - 13 | 1426 | 69 | ||
AK167975.1 | - | 12 - 13 | 1424 | 69 | ||
AK169325.1 | - | 12 - 13 | 1420 | 69 | ||
AK169342.1 | - | 12 - 13 | 1420 | 69 | ||
BC010810.1 | - | 12 - 13 | 1410 | 4241697 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21078344 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1992525 | AK032081.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK169342.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK167975.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK169325.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK158990.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK147110.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK169342.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK167975.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK169325.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK158990.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P504302 | AK147110.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661335 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112299 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520964 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241430 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529839 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801338 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 374876 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131648 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152326 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144631 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93135 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 58081 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123458 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138136 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147210 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK169342.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK167975.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK169325.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK158990.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK147110.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK169342.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK167975.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK169325.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK158990.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P504302 | AK147110.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078386 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078378 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078361 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078372 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078348 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078387 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078363 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078370 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078346 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078373 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078375 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078383 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078376 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078355 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078381 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078385 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078352 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078379 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078351 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078366 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078357 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078364 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078353 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078349 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21078344 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147210 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123458 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138136 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00326845 | - | - |
Whole Mouse Genome G4122A | A_52_P504302 | AK169342.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P504302 | AK167975.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P504302 | AK169325.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P504302 | AK158990.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P504302 | AK147110.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P504302 | AK169342.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P504302 | AK167975.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P504302 | AK169325.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P504302 | AK158990.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P504302 | AK147110.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P569651 | AK169342.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P569651 | BC010810.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P569651 | AK167975.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P569651 | AK169325.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P569651 | AK161291.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119941 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137619 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138136 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147210 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123458 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01906136 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01700973 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01906139 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01700977 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21078344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21078398 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01232822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078344 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078348 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01700972 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078373 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078396 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078366 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01906139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078381 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01700975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01299818 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01700959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078349 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078398 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01700969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02268294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01700966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01700971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21078394 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10470273 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470274 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470279 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470280 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470276 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470270 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470275 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470277 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470271 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470282 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470278 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470272 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470269 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470281 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10481012 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424662_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424661_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10470268 | - | - |
Mouse Genome 430 2.0 Array | 1424662_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1424662_at | - | - |
Mouse Genome 430 2.0 Array | 1424661_at | - | - |
Mouse Genome 430 2.0 Array | 1424661_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66865_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66865_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200002865.mm.1 | TC0200000450.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200002865.mm.1 | NM_173180.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200002865.mm.1 | NM_173180.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200002866.mm.1 | TC0200000450.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200002866.mm.1 | NM_173180.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200002866.mm.1 | NM_173180.3 | Refseq |
Mouse Expression Set 430 | 1424661_at | - | - |
Mouse Expression Set 430 | 1424662_at | - | - |
Mouse Expression Set 430 | 1424662_PM_at | - | - |
Mouse Expression Set 430 | 1424661_PM_at | - | - |
Murine Genome U74v2 Set | 96848_at | - | - |
Murine Genome U74v2 Set | 96848_PM_at | - | - |
Murine Genome U74v2 Set | 168492_at | - | - |
Murine Genome U74v2 Set | 134130_at | - | - |
Murine Genome U74v2 Set | 168492_PM_at | - | - |
Murine Genome U74v2 Set | 134130_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1424662_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1424662_at | - | - |
Mouse Genome 430A 2.0 Array | 1424661_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1424661_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424662_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424662_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424661_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424661_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4280132 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6840408 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006840408 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004280132 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1251648 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227913 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2643846 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1251648 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2643846 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1227913 | - | - |
mouse-6 v1.1 (Illumina) | scl0066865.1_100-S | - | - |
mouse-6 v1.1 (Illumina) | scl066865.9_12-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4280132 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6840408 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl066865.9_12-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066865.1_100-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840408 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280132 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1227913 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227913 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066865.9_12-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066865.1_100-S | - | - |
