Assay Details
Gene Symbol: P3h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173379.2 | NP_775555.1 | 2 - 3 | 717 | 131 | |
GenBank mRNA | AJ430350.1 | - | 2 - 3 | 718 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
