Assay Details
Gene Symbol: Gpr171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173398.3 | NP_775574.1 | 1 - 2 | 237 | 147 | |
GenBank mRNA | AK041381.1 | - | 1 - 2 | 210 | 146 | |
AK041582.1 | - | 1 - 2 | 235 | 145 | ||
AK079674.1 | - | 1 - 2 | 238 | 147 | ||
AK083323.1 | - | 1 - 2 | 235 | 146 | ||
AK089252.1 | - | 1 - 2 | 235 | 150 | ||
AK089287.1 | - | 1 - 2 | 189 | 148 | ||
AK154725.1 | - | 1 - 2 | 235 | 147 | ||
BC024054.1 | - | 2 - 3 | 5068166 | 138 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK041381.1,AK041582.1,AK089252.1,AK089287.1,AK154725.1,BC024054.1,NM_173398.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
