Assay Details
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174991.3 | NP_778156.2 | 28 - 29 | 5301 | 59 | |
GenBank mRNA | AK046831.1 | - | 12 - 13 | 1819 | 59 | |
AK147456.1 | - | 28 - 29 | 5286 | 59 | ||
AK147459.1 | - | 28 - 29 | 5254 | 59 | ||
AK147494.1 | - | 28 - 29 | 5088 | 59 | ||
AK147600.1 | - | 27 - 28 | 4977 | 59 | ||
AK147607.1 | - | 28 - 29 | 5305 | 59 | ||
AK220481.1 | - | 28 - 29 | 4695 | 59 | ||
AY168408.1 | - | 28 - 29 | 4813 | 59 | ||
BC036963.1 | - | 10 - 11 | 1479 | 59 | ||
BC059857.1 | - | 7 - 8 | 1231 | 59 | ||
BC115835.1 | - | 4 - 5 | 1421 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
