Assay Details
Gene Symbol: Rab11fip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175543.3 | NP_780752.1 | 4 - 5 | 623 | 98 | |
GenBank mRNA | AK046734.1 | - | 4 - 5 | 625 | 98 | |
AK147219.1 | - | 4 - 5 | 694 | 98 | ||
AK147344.1 | - | 4 - 5 | 819 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
