Assay Details
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175628.3 | NP_783327.2 | 28 - 29 | 3654 | 99 | |
GenBank mRNA | AY185125.1 | - | 28 - 29 | 3600 | 99 | |
BC023844.1 | - | 18 - 19 | 2316 | 99 | ||
BC072642.1 | - | 28 - 29 | 3594 | 6839761 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
