Assay Details
Gene Symbol: Ptk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175168.4 | NP_780377.1 | 6 - 7 | 1118 | 59 | |
GenBank mRNA | AF531873.1 | - | 6 - 7 | 968 | 59 | |
AK053044.1 | - | 6 - 7 | 1051 | 59 | ||
AK163135.1 | - | 6 - 7 | 1051 | 59 | ||
BC076578.1 | - | 6 - 7 | 1118 | 30619280 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
