Assay Details
Gene Symbol: Nell1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037906.2 | NP_001032995.1 | 5 - 6 | 638 | 79 | |
GenBank mRNA | AK032845.1 | - | 5 - 6 | 920 | 79 | |
AY622226.1 | - | 5 - 6 | 638 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK035921.1 | - | 1127 | Chr. - - [-] on Build | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
