Assay Details
Gene Symbol: Uckl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026765.3 | NP_081041.2 | 3 - 4 | 419 | 96 | |
GenBank mRNA | AK144803.1 | - | 3 - 4 | 425 | 96 | |
BC016535.1 | - | 3 - 4 | 419 | 2655873 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
