Assay Details
Gene Symbol: Ppp1r21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028658.4 | NP_082934.3 | 16 - 17 | 1828 | 131 | |
GenBank mRNA | AK028185.1 | - | 16 - 17 | 1692 | 131 | |
AK155094.1 | - | 16 - 17 | 1660 | 131 | ||
AK170254.1 | - | 16 - 17 | 1716 | 131 | ||
BC137673.1 | - | 16 - 17 | 1768 | 8860692 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
