Assay Details
Gene Symbol: Ppp1r21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028658.4 | NP_082934.3 | 8 - 9 | 885 | 62 | |
GenBank mRNA | AK028185.1 | - | 8 - 9 | 749 | 62 | |
BC137673.1 | - | 8 - 9 | 825 | 8860692 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
