Assay Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146043.2 | NP_001139515.1 | 12 - 13 | 1194 | 82 | |
NM_001271463.1 | NP_001258392.1 | 12 - 13 | 1194 | 82 | ||
NM_001271464.1 | NP_001258393.1 | 11 - 12 | 1113 | 82 | ||
NM_001271465.1 | NP_001258394.1 | 12 - 13 | 1221 | 82 | ||
NM_177266.5 | NP_796240.3 | 12 - 13 | 1227 | 82 | ||
GenBank mRNA | AK028450.1 | - | 12 - 13 | 1180 | 82 | |
AK035819.1 | - | 12 - 13 | 1133 | 82 | ||
AK044371.1 | - | 11 - 12 | 1114 | 82 | ||
AK083441.1 | - | 12 - 13 | 1162 | 83 | ||
AK161797.1 | - | 12 - 13 | 1130 | 82 | ||
AK165871.1 | - | 12 - 13 | 1188 | 82 | ||
BC057062.1 | - | 1 - 2 | 1005 | 82 | ||
BC138233.1 | - | 12 - 13 | 1125 | 8861252 | 82 | |
BC145208.1 | - | 12 - 13 | 1125 | 9053759 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
