Assay Details
Gene Symbol: Ercc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007949.4 | NP_031975.2 | 19 - 20 | 1850 | 113 | |
GenBank mRNA | AK004652.1 | - | 15 - 16 | 1502 | 113 | |
AK082323.1 | - | 12 - 13 | 1104 | 113 | ||
AK082761.1 | - | 19 - 20 | 1851 | 113 | ||
AK209753.1 | - | 4 - 5 | 314 | 113 | ||
BC034517.1 | - | 18 - 19 | 1786 | 4458839 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
