Assay Details
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133847.3 | NP_598608.2 | 1 - 2 | 266 | 65 | |
GenBank mRNA | AK035622.1 | - | 1 - 2 | 228 | 65 | |
AK083541.1 | - | 1 - 2 | 266 | 65 | ||
BC116320.2 | - | 1 - 2 | 262 | 40096002 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK083541.1, BC116320.2, NM_133847.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
