Assay Details
Gene Symbol: Oas1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133893.3 | NP_598654.1 | 5 - 6 | 1121 | 153 | |
GenBank mRNA | AB067532.1 | - | 5 - 6 | 1075 | 153 | |
AK054465.1 | - | 5 - 6 | 1089 | 153 | ||
AK135783.1 | - | 4 - 5 | 635 | 153 | ||
AK136001.1 | - | 5 - 6 | 1089 | 153 | ||
AY055829.1 | - | 5 - 6 | 1083 | 153 | ||
BC052835.1 | - | 5 - 6 | 1085 | 30048758 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989071 | AY055829.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989071 | BC052835.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989071 | AB067532.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989071 | AK054465.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989071 | AK135783.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51750 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715769 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154163 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52365 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146527 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989071 | AY055829.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989071 | BC052835.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989071 | AB067532.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989071 | AK054465.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989071 | AK135783.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183025 | AY055829.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183025 | BC052835.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183025 | AB067532.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183025 | AK054465.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183025 | AK135783.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041555 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041544 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041543 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041550 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146527 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05720441 | - | - |
Whole Mouse Genome G4122A | A_51_P183025 | AY055829.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183025 | BC052835.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183025 | AB067532.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183025 | AK054465.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183025 | AK135783.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133680 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04140297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041622 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04266754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04139023 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04303933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041544 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04140296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041564 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041616 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041627 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041555 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041584 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04418562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041559 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04266753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041585 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04266751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041553 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041573 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041572 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041527 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10525181 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525182 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525184 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525183 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10525180 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10525179 | - | - |
Mouse Genome 430 2.0 Array | 1416847_s_at | - | - |
Mouse Genome 430 2.0 Array | 1416847_PM_s_at | - | - |
Mouse Expression Set 430 | 1416847_PM_s_at | - | - |
Mouse Expression Set 430 | 1416847_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1416847_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1416847_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2100162 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002100162 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2737613 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2737613 | - | - |
mouse-6 v1.1 (Illumina) | scl27285.6.1_21-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2100162 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl27285.6.1_21-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100162 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2737613 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2929966 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2737613 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2929966 | - | - |
