Assay Details
Gene Symbol: Actg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009610.2 | NP_033740.2 | 7 - 8 | 911 | 150 | |
GenBank mRNA | AK146631.1 | - | 7 - 8 | 884 | 150 | |
AK168649.1 | - | 7 - 8 | 884 | 150 | ||
BC002042.1 | - | 7 - 8 | 892 | 3591323 | 150 | |
BC049611.1 | - | 7 - 8 | 891 | 6773765 | 150 | |
M26689.1 | - | 6 - 7 | 840 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963807 | BC002042.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963807 | NM_009610.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915431 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789208 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48551 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367781 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364327 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281890 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 222699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 466096 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655470 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910010 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739544 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811413 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851855 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19585 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156553 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118056 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163197 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963807 | BC002042.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963807 | NM_009610.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241269 | BC049611.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241269 | M26689.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241269 | AK146631.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241269 | AK168649.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241269 | NM_009610.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594070 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594082 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594085 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594071 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594087 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594090 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594097 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594075 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594077 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24594078 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163197 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05961880 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118056 | - | - |
Whole Mouse Genome G4122A | A_51_P241269 | BC049611.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P241269 | M26689.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P241269 | AK146631.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P241269 | AK168649.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P241269 | NM_009610.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100140 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102515 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118056 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163197 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05557606 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05557621 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05557624 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05410120 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05557613 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05410121 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05084663 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05557610 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05557607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24594071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24593995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593967 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05224066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05698419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05292713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05121899 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05224063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05201671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05410120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05084663 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05672060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594077 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05611829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594090 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594033 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05557610 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593973 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593972 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05492068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593988 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594100 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05611828 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05741303 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05329957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24593987 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05557621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24594030 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10545711 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545708 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545714 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545712 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545715 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545717 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545709 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545713 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545710 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545719 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545718 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10545716 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422340_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10545707 | - | - |
Mouse Genome 430 2.0 Array | 1422340_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1422340_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11468_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11468_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600019972.mm.1 | TC0600002574.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600019972.mm.1 | NM_009610.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600019972.mm.1 | NM_009610.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600019971.mm.1 | TC0600002574.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600019971.mm.1 | NM_009610.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600019971.mm.1 | NM_009610.2 | Refseq |
Mouse Expression Set 430 | 1422340_a_at | - | - |
Mouse Expression Set 430 | 1422340_PM_a_at | - | - |
Murine Genome U74v2 Set | 93102_PM_f_at | - | - |
Murine Genome U74v2 Set | 93102_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1422340_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1422340_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422340_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422340_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 912820 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4780180 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4780180 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004780180 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2609642 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2609642 | - | - |
mouse-6 v1.1 (Illumina) | scl28794.10.1_120-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4780180 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl28794.10.1_120-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780180 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2839313 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2839313 | - | - |
