Assay Details
Gene Symbol: Iqca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016353.1 | - | 6 - 7 | 1151 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021275 | AK139726.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021275 | NM_029122.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85062 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845452 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329512 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129672 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101671 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021275 | AK139726.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021275 | NM_029122.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129672 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00129945 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101671 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100230 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129672 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101671 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434216 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434223 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434056 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00242061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00655696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00758048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01048528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434122 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00242048 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00417491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00056680 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00242043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434206 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00696083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00061544 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434159 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00504807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00056681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434187 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00902037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434110 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00892718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00417489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434328 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00738195 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434116 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00107597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00242050 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434213 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434165 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434323 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434198 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434312 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00371117 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00242047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00902039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434067 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434208 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434218 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434095 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00559784 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434320 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00812784 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00922643 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434321 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434038 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00620624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434160 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00812788 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00902032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00559786 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00696091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434310 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434112 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434162 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434205 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434102 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00660317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00902035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00696087 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00441236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434058 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00242063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00902034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434330 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00696095 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00559789 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01042246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20434037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00797852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00797849 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10356519 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10356515 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10356517 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10356518 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10356516 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435992_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432060_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10356510 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10356514 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10356512 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10356498 | - | - |
Mouse Genome 430 2.0 Array | 1435992_at | - | - |
Mouse Genome 430 2.0 Array | 1435992_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432060_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432060_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74918_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74918_PM_at | - | - |
Mouse Expression Set 430 | 1435992_PM_at | - | - |
Mouse Expression Set 430 | 1435992_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432060_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432060_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435992_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435992_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 631791 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 101780093 | - | - |
MouseWG-6 v2 Expression BeadChip | 4810433 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4810433 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004810433 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2654737 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2654737 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1248198 | - | - |
mouse-6 v1.1 (Illumina) | scl0074918.2_57-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4810433 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0074918.2_57-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810433 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2654736 | - | - |
