Assay Details
Gene Symbol: Dhx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026987.2 | NP_081263.2 | 12 - 13 | 2153 | 78 | |
GenBank mRNA | AK122314.2 | - | 12 - 13 | 2088 | 78 | |
BC009147.1 | - | 12 - 13 | 2112 | 2647184 | 78 | |
BC138204.1 | - | 12 - 13 | 2092 | 8861223 | 78 | |
BC138205.1 | - | 12 - 13 | 2092 | 8861224 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
