Assay Details
Gene Symbol: Ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172073.1 | NP_001165544.1 | 4 - 5 | 553 | 69 | |
NM_028209.2 | NP_082485.1 | 4 - 5 | 552 | 69 | ||
GenBank mRNA | AK005923.1 | - | 4 - 5 | 554 | 69 | |
AK012652.1 | - | 4 - 5 | 498 | 69 | ||
AK088684.1 | - | 4 - 5 | 514 | 69 | ||
AK133931.1 | - | 4 - 5 | 517 | 69 | ||
BC025435.1 | - | 4 - 5 | 501 | 5004463 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
