Assay Details
Gene Symbol: Sf3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030109.2 | NP_084385.2 | 12 - 13 | 1641 | 90 | |
GenBank mRNA | AK046493.1 | - | 2 - 3 | 306 | 90 | |
AK142136.1 | - | 12 - 13 | 1611 | 90 | ||
AK146063.1 | - | 12 - 13 | 1611 | 90 | ||
AK146540.1 | - | 12 - 13 | 1606 | 90 | ||
AK146864.1 | - | 12 - 13 | 1610 | 90 | ||
AK147687.1 | - | 12 - 13 | 1609 | 90 | ||
AK149776.1 | - | 12 - 13 | 1612 | 90 | ||
AK150961.1 | - | 12 - 13 | 1598 | 90 | ||
AK151354.1 | - | 12 - 13 | 1588 | 90 | ||
AK152327.1 | - | 12 - 13 | 1593 | 90 | ||
AK152965.1 | - | 12 - 13 | 1598 | 90 | ||
AK159893.1 | - | 12 - 13 | 1600 | 90 | ||
AK210738.1 | - | 1 - 2 | 149 | 90 | ||
AK213001.1 | - | 1 - 2 | 149 | 90 | ||
AK216291.1 | - | 1 - 2 | 149 | 90 | ||
AK219963.1 | - | 1 - 2 | 149 | 90 | ||
BC029745.1 | - | 12 - 13 | 1575 | 90 | ||
BC045207.1 | - | 12 - 13 | 1610 | 90 | ||
BC049118.1 | - | 12 - 13 | 1634 | 6812422 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
