Assay Details
Gene Symbol: Dgkz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166597.1 | NP_001160069.1 | 4 - 5 | 1079 | 95 | |
NM_138306.2 | NP_612179.2 | 3 - 4 | 478 | 95 | ||
GenBank mRNA | AK087503.1 | - | 4 - 5 | 1080 | 95 | |
AK201965.1 | - | 1 - 2 | 50 | 95 | ||
BC016217.1 | - | 3 - 4 | 1425 | 95 | ||
BC049228.1 | - | 3 - 4 | 514 | 3668704 | 95 | |
BY733951.1 | - | 3 - 4 | 479 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
