Assay Details
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253831.1 | NP_001240760.1 | 13 - 14 | 1325 | 100 | |
NM_001253834.1 | NP_001240763.1 | 14 - 15 | 1416 | 100 | ||
NM_001253836.1 | NP_001240765.1 | 13 - 14 | 1348 | 100 | ||
NM_175025.4 | NP_778190.3 | 14 - 15 | 1531 | 100 | ||
GenBank mRNA | AJ551270.1 | - | 12 - 13 | 1113 | 100 | |
AK028393.1 | - | 13 - 14 | 1294 | 100 | ||
AK133703.1 | - | 13 - 14 | 1326 | 100 | ||
AK159918.1 | - | 14 - 15 | 100 | |||
AK163049.1 | - | 14 - 15 | 1377 | 100 | ||
AK173154.1 | - | 12 - 13 | 1104 | 100 | ||
BC043091.1 | - | 14 - 15 | 1497 | 6405251 | 100 | |
BC043706.1 | - | 14 - 15 | 1477 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
