Assay Details
Gene Symbol: Oas1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF480417.1 | - | 7 | 1946 | 90 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998979 | AF480417.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998979 | NM_011852.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998977 | AF480417.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998977 | NM_011852.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798192 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615313 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61303 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117059 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57917 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147619 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998977 | AF480417.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998977 | NM_011852.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998979 | AF480417.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998979 | NM_011852.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041405 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041391 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041418 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041389 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041408 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041403 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041407 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24041381 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05720331 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147619 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01204631 | - | - |
Whole Mouse Genome G4122A | A_51_P355267 | X58077.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P355267 | AF480417.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P355267 | NM_011852.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134752 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138405 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139839 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105132 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101184 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147619 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04246505 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04264207 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04246508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24041283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041395 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041381 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041418 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04335121 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041414 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04460852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041408 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041407 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04119582 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24041402 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10533253 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10533254 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10533251 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10533250 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10533247 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10533249 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10533248 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10533255 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10533252 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10533246 | - | - |
Murine Genome U74v2 Set | 102717_PM_at | - | - |
Murine Genome U74v2 Set | 102717_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 394201 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2450280 | - | - |
MouseWG-6 v2 Expression BeadChip | 3990070 | - | - |
MouseWG-6 v2 Expression BeadChip | 3170451 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3170451 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3990070 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1940458 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2450280 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002450280 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003990070 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001940458 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003170451 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255890 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706310 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2628822 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253808 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2624469 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2628822 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2624469 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1253808 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2706310 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1255890 | - | - |
mouse-6 v1.1 (Illumina) | scl0023960.2_17-S | - | - |
mouse-6 v1.1 (Illumina) | scl023960.6_30-S | - | - |
mouse-6 v1.1 (Illumina) | scl0246730.3_14-S | - | - |
mouse-6 v1.1 (Illumina) | scl0023960.2_325-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1940458 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2450280 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3170451 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3990070 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl023960.6_30-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0246730.3_14-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0023960.2_325-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0023960.2_17-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2450280 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170451 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3990070 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940458 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2816196 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2628822 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2624469 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1253808 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253808 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2624469 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2628822 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2816196 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0246730.3_14-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl023960.6_30-S | - | - |
