Assay Details
Gene Symbol: Ifna13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177347.2 | NP_796321.1 | 1 | 747 | 125 | |
GenBank mRNA | AY190047.1 | - | 1 | 747 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
defense response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
defense response to virus
cytokine activity
cytokine receptor binding
type I interferon receptor binding
adaptive immune response
T cell activation involved in immune response
natural killer cell activation involved in immune response
defense response
humoral immune response
cytokine-mediated signaling pathway
B cell differentiation
positive regulation of peptidyl-serine phosphorylation of STAT protein
B cell proliferation
response to exogenous dsRNA
defense response to virus
cytokine activity
cytokine receptor binding
type I interferon receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166020 | NM_177347.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166020 | AY190047.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166020 | BC120724.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166020 | BC120750.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898878 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170643 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166020 | NM_177347.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166020 | AY190047.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166020 | BC120724.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166020 | BC120750.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P355382 | NM_177347.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P355382 | AY190047.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P355382 | BC120724.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P355382 | BC120750.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091548 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091524 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091533 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091550 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091552 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091538 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091549 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091528 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091540 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091525 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091535 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091536 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091542 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170643 | - | - |
Whole Mouse Genome G4122A | A_51_P355382 | NM_177347.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P355382 | AY190047.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P355382 | BC120724.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P355382 | BC120750.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170643 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03554111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091540 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091555 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091548 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03679218 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03554111 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091516 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03457150 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03457148 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091552 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091553 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091537 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03457149 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23091550 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10514312 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10514311 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10514310 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10514309 | - | - |
Affymetrix Mouse Clariom S Array | PSR0400024326.mm.1 | TC0400003009.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400024326.mm.1 | NM_177347.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400024326.mm.1 | NM_177347.2 | Refseq |
Mouse Expression Set 430 | 1450593_PM_at | - | - |
Mouse Expression Set 430 | 1422403_at | - | - |
Mouse Expression Set 430 | 1422403_PM_at | - | - |
Mouse Expression Set 430 | 1422404_PM_x_at | - | - |
Mouse Expression Set 430 | 1422404_x_at | - | - |
Mouse Expression Set 430 | 1450593_at | - | - |
Murine Genome U74v2 Set | 102149_f_at | - | - |
Murine Genome U74v2 Set | 101619_at | - | - |
Murine Genome U74v2 Set | 102149_PM_f_at | - | - |
Murine Genome U74v2 Set | 101619_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422403_at | - | - |
Mouse Genome 430A 2.0 Array | 1450593_at | - | - |
Mouse Genome 430A 2.0 Array | 1450593_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422404_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1422404_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1422403_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 363499 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1980086 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106900537 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001980086 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2641897 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1220308 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2641897 | - | - |
mouse-6 v1.1 (Illumina) | scl24129.1.1_238-S | - | - |
mouse-6 v1.1 (Illumina) | GI_31340904-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1980086 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106900537 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl24129.1.1_238-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980086 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106900537 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2641897 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2641897 | - | - |
