Assay Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290413.1 | NP_001277342.1 | 7 - 8 | 811 | 57 | |
NM_009422.3 | NP_033448.2 | 7 - 8 | 790 | 57 | ||
GenBank mRNA | AF027570.1 | - | 6 - 7 | 702 | 57 | |
AK038136.1 | - | 7 - 8 | 739 | 57 | ||
AK052934.1 | - | 7 - 8 | 739 | 57 | ||
AK151653.1 | - | 7 - 8 | 728 | 57 | ||
AK152174.1 | - | 7 - 8 | 758 | 57 | ||
AK159581.1 | - | 7 - 8 | 760 | 57 | ||
AK159585.1 | - | 737 | 57 | |||
BC003801.1 | - | 6 - 7 | 716 | 3492839 | 57 | |
BC060625.1 | - | 5 - 6 | 2747 | 57 | ||
L35303.1 | - | 7 - 8 | 727 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
