Assay Details
Gene Symbol: Adgre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010130.4 | NP_034260.1 | 15 - 16 | 1966 | 71 | |
GenBank mRNA | AK133202.1 | - | 15 - 16 | 1942 | 71 | |
AK149898.1 | - | 15 - 16 | 1951 | 71 | ||
AK151303.1 | - | 15 - 16 | 1938 | 71 | ||
AK151682.1 | - | 15 - 16 | 1958 | 71 | ||
AK155862.1 | - | 15 - 16 | 1917 | 71 | ||
BC075688.1 | - | 15 - 16 | 1938 | 30609706 | 71 | |
DQ167573.1 | - | 1 - 2 | 150 | 71 | ||
U66888.1 | - | 15 - 16 | 1953 | 71 | ||
X93328.1 | - | 15 - 16 | 1962 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
