Assay Details
Gene Symbol: Pip4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054051.1 | NP_473392.1 | 3 - 4 | 448 | 59 | |
GenBank mRNA | AK089414.1 | - | 3 - 4 | 438 | 59 | |
AK146559.1 | - | 3 - 4 | 498 | 59 | ||
AK170392.1 | - | 3 - 4 | 375 | 59 | ||
AK171497.1 | - | 3 - 4 | 438 | 59 | ||
AY050219.1 | - | 3 - 4 | 277 | 59 | ||
BC047282.1 | - | 3 - 4 | 448 | 6491155 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
