Assay Details
Gene Symbol: Hadha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178878.2 | NP_849209.1 | 10 - 11 | 1008 | 63 | |
GenBank mRNA | AK029017.1 | - | 10 - 11 | 1009 | 63 | |
AK150188.1 | - | 10 - 11 | 1006 | 63 | ||
AK170478.1 | - | 10 - 11 | 1009 | 63 | ||
AK170683.1 | - | 10 - 11 | 1009 | 63 | ||
AK171469.1 | - | 10 - 11 | 1009 | 63 | ||
AK179416.1 | - | 2 - 3 | 168 | 63 | ||
AK188264.1 | - | 2 - 3 | 168 | 63 | ||
AK217494.1 | - | 2 - 3 | 169 | 63 | ||
BC037009.1 | - | 10 - 11 | 996 | 5134605 | 63 | |
BC046978.1 | - | 10 - 11 | 982 | 6315204 | 63 | |
BC058569.1 | - | 10 - 11 | 1004 | 6468514 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
