Assay Details
Gene Symbol: Zfp598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183149.1 | NP_898972.1 | 2 - 3 | 334 | 57 | |
GenBank mRNA | AK131127.1 | - | 2 - 3 | 353 | 57 | |
BC024690.1 | - | 2 - 3 | 128 | 57 | ||
BC050859.1 | - | 2 - 3 | 334 | 6328878 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
