Assay Details
Gene Symbol: Slc9c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198106.4 | NP_932774.3 | 18 - 19 | 2585 | 143 | |
GenBank mRNA | AK131702.1 | - | 1 - 2 | 126 | 143 | |
AY368685.1 | - | 18 - 19 | 2545 | 143 | ||
BC145031.1 | - | 2192 | 143 | |||
BC145032.1 | - | 2407 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
