Assay Details
Gene Symbol: Ang | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161731.2 | NP_001155203.1 | 2 | 231 | 86 | |
NM_007447.3 | NP_031473.1 | 2 | 528 | 86 | ||
GenBank mRNA | AK159966.1 | - | 2 | 529 | 86 | |
AK171251.1 | - | 2 | 159 | 86 | ||
BC055355.1 | - | 2 | 115 | 5136641 | 86 | |
BF721354.1 | - | 2 | 104 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
response to hypoxia
positive regulation of endothelial cell proliferation
diacylglycerol biosynthetic process
activation of phospholipase C activity
multicellular organism development
central nervous system development
rRNA transcription
response to hormone
cell migration
negative regulation of translation
cell differentiation
activation of protein kinase B activity
activation of phospholipase A2 activity
positive regulation of phosphorylation
negative regulation of smooth muscle cell proliferation
positive regulation of protein secretion
nucleic acid phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis
nucleic acid binding
DNA binding
RNA binding
actin binding
nuclease activity
endonuclease activity
ribonuclease activity
signaling receptor binding
copper ion binding
heparin binding
hydrolase activity
peptide binding
protein homodimerization activity
angiogenesis
response to hypoxia
positive regulation of endothelial cell proliferation
diacylglycerol biosynthetic process
activation of phospholipase C activity
multicellular organism development
central nervous system development
rRNA transcription
response to hormone
cell migration
negative regulation of translation
cell differentiation
activation of protein kinase B activity
activation of phospholipase A2 activity
positive regulation of phosphorylation
negative regulation of smooth muscle cell proliferation
positive regulation of protein secretion
nucleic acid phosphodiester bond hydrolysis
RNA phosphodiester bond hydrolysis
nucleic acid binding
DNA binding
RNA binding
actin binding
nuclease activity
endonuclease activity
ribonuclease activity
signaling receptor binding
copper ion binding
heparin binding
hydrolase activity
peptide binding
protein homodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | NM_007447.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | AK159966.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | AK171251.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | BC055355.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | BF721354.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | NM_007447.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | AK159966.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | AK171251.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | BC055355.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P391159 | BF721354.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314332 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889641 | AK017333.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889641 | AK028741.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889641 | AK054164.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889641 | BC076614.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889641 | BC079647.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889641 | NM_181815.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295164 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224426 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17263 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151128 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | NM_007447.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | AK159966.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | AK171251.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | BC055355.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | BF721354.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | NM_007447.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | AK159966.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | AK171251.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | BC055355.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P391159 | BF721354.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | NM_007447.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | AK159966.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | AK171251.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | BC055355.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | BF721354.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | NM_007447.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | AK159966.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | AK171251.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | BC055355.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P391159 | BF721354.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29600072 | - | - |
Whole Mouse Genome G4122A | A_51_P391159 | NM_007447.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391159 | AK159966.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391159 | AK171251.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391159 | BC055355.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391159 | BF721354.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391159 | NM_007447.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391159 | AK159966.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391159 | AK171251.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391159 | BC055355.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P391159 | BF721354.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127600 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108481 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11206038 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11206037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29600096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11657583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600072 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11657585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600103 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600091 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600106 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600096 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29600084 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11406545 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11206037 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10414540 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10414538 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10414539 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10414541 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438936_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450717_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438937_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10414537 | - | - |
Mouse Genome 430 2.0 Array | 1438937_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1438936_s_at | - | - |
Mouse Genome 430 2.0 Array | 1450717_at | - | - |
Mouse Genome 430 2.0 Array | 1438936_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1438937_x_at | - | - |
Mouse Genome 430 2.0 Array | 1450717_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11727_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11727_PM_at | - | - |
Mouse Expression Set 430 | 1450717_PM_at | - | - |
Mouse Expression Set 430 | 1450717_at | - | - |
Murine Genome U74v2 Set | 94392_PM_f_at | - | - |
Murine Genome U74v2 Set | 94392_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1450717_at | - | - |
Mouse Genome 430A 2.0 Array | 1450717_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450717_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450717_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438937_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438936_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438936_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438937_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 370353 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3990368 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3990368 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003990368 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773844 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2773844 | - | - |
mouse-6 v1.1 (Illumina) | scl011727.2_300-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3990368 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl011727.2_300-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3990368 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2875251 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2683113 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2875251 | - | - |
