Assay Details
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009760.4 | NP_033890.1 | 2 - 3 | 291 | 125 | |
GenBank mRNA | AF041054.1 | - | 2 - 3 | 261 | 125 | |
AK014223.1 | - | 2 - 3 | 292 | 125 | ||
AK075943.1 | - | 2 - 3 | 292 | 125 | ||
AK152610.1 | - | 2 - 3 | 287 | 125 | ||
BC033278.1 | - | 2 - 3 | 199 | 125 | ||
BC046603.1 | - | 2 - 3 | 281 | 6390074 | 125 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
